Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LPAT4
Entrez Gene ID
843840
Gene Name
lysophosphatidyl acyltransferase 4
Gene Aliases
AT1G75020, F25A4.2, F25A4_2, lysophosphatidyl acyltransferase 4
Chromosome Location
Chr.1: 28170752 - 28173678 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_106159.3 NP_565098.1 2 - 3 875 69
NM_202415.2 NP_974144.1 1 - 2 1599 69
GenBank mRNA AY084709.1 - 2 - 3 701 69
AY099547.1 - 2 - 3 701 69
BT001236.1 - 1 - 2 572 69

View Full Product Details