Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
JAC1
Entrez Gene ID
843847
Gene Name
J-domain protein required for chloroplast accumulation response 1
Gene Aliases
AT1G75100, F9E10.5, F9E10_5, J-domain protein required for chloroplast accumulation response 1
Chromosome Location
Chr.1: 28190740 - 28194068 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_106166.4 NP_565101.1 5 - 6 1894 80
GenBank mRNA AB158477.1 - 5 - 6 1608 80
AY057504.1 - 5 - 6 1626 80
AY103303.1 - 5 - 6 1595 80

View Full Product Details