Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RRA1
Entrez Gene ID
843849
Gene Name
Nucleotide-diphospho-sugar transferase family protein
Gene Aliases
AT1G75120, F9E10.3, F9E10_3, REDUCED RESIDUAL ARABINOSE 1
Chromosome Location
Chr.1: 28196857 - 28198945 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_106168.3 NP_177648.1 1 - 2 426 67
GenBank mRNA BT026432.1 - 1 - 2 137 67
KJ138931.1 - 1 - 2 137 67

View Full Product Details