Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- BBX21
- Entrez Gene ID
-
843890
- Gene Name
- salt tolerance homolog2
- Gene Aliases
-
AT1G75540, AtBBX21, B BOX 21, B-box domain protein 21, F10A5.24, F10A5_24, LHUS, STH2, long hypocotyl under shade, salt tolerance homolog2
- Chromosome Location
-
Chr.1: 28365824 - 28367738 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_106207.4 |
NP_177686.1 |
2 - 3 |
852 |
81 |
GenBank mRNA |
AB493536.1 |
- |
2 - 3 |
617 |
81 |
View Full Product Details