Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- XRN3
- Entrez Gene ID
-
843900
- Gene Name
- 5'-3' exoribonuclease 3
- Gene Aliases
-
5'-3' EXORIBONUCLEASE 3, 5'-3' exoribonuclease 3, AT1G75660, AtXRN3, F10A5.15, F10A5_15
- Chromosome Location
-
Chr.1: 28408134 - 28415118 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_106217.3 |
NP_565114.1 |
7 - 8 |
1066 |
92 |
View Full Product Details