Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CLV1
- Entrez Gene ID
-
843915
- Gene Name
- Leucine-rich receptor-like protein kinase family protein
- Gene Aliases
-
AT1G75820, ATCLV1, CLAVATA 1, CLV1 RECEPTOR KINASE PRECURSOR, FAS3, FASCIATA 3, FLO5, FLOWER DEVELOPMENT 5, T4O12.5, T4O12_5
- Chromosome Location
-
Chr.1: 28463344 - 28466971 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_106232.4 |
NP_177710.1 |
1 - 2 |
2906 |
71 |
GenBank mRNA |
FJ708682.1 |
- |
1 - 2 |
2587 |
71 |
View Full Product Details