Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CPK29
Entrez Gene ID
843936
Gene Name
calcium-dependent protein kinase 29
Gene Aliases
AT1G76040, T4O12.25, T4O12_25, calcium-dependent protein kinase 29
Chromosome Location
Chr.1: 28537593 - 28540707 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001334707.1 NP_001319387.1 2 - 3 1412 72
NM_001334708.1 NP_001320822.1 2 - 3 1094 72
NM_106253.4 NP_177731.2 1 - 2 378 72
NM_202421.3 NP_974150.2 2 - 3 1094 72
GenBank mRNA AY093022.1 - 1 - 2 308 72
BT000401.1 - 1 - 2 230 72

View Full Product Details