Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYCB2;4
Entrez Gene ID
843964
Gene Name
CYCLIN B2;4
Gene Aliases
AT1G76310, CYCLIN B2;4, F15M4.19
Chromosome Location
Chr.1: 28627694 - 28630789 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001334719.1 NP_001323250.1 3 - 4 497 84
NM_001334720.1 NP_001323249.1 3 - 4 376 84
NM_001334721.1 NP_001323251.1 3 - 4 376 84
NM_106281.6 NP_177758.2 3 - 4 619 84

View Full Product Details