Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PIN6
Entrez Gene ID
844046
Gene Name
Auxin efflux carrier family protein
Gene Aliases
AT1G77110, F22K20.18, F22K20_18, PIN-FORMED 6
Chromosome Location
Chr.1: 28970714 - 28974805 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001334766.1 NP_001323018.1 1 - 2 566 71
NM_106361.4 NP_177836.1 1 - 2 566 71

View Full Product Details