Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AAP3
Entrez Gene ID
844074
Gene Name
amino acid permease 3
Gene Aliases
AT1G77380, ATAAP3, F2P24.9, F2P24_9, amino acid permease 3
Chromosome Location
Chr.1: 29074879 - 29077390 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_106387.3 NP_177862.1 1 - 2 232 82
GenBank mRNA AY099788.1 - 1 - 2 232 82
AY128905.1 - 1 - 2 94 82
X77499.1 - - 181 82

View Full Product Details