Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LACS9
Entrez Gene ID
844094
Gene Name
long chain acyl-CoA synthetase 9
Gene Aliases
AT1G77590, T5M16.18, T5M16_18, long chain acyl-CoA synthetase 9
Chromosome Location
Chr.1: 29148160 - 29152395 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001334788.1 NP_001321320.1 10 - 11 2261 108
NM_106407.7 NP_177882.1 11 - 12 2188 108
GenBank mRNA AF503759.1 - 10 - 11 1913 108

View Full Product Details