The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_106421.5 | NP_565160.3 | 1 - 2 | 402 | 67 |
GenBank mRNA | AK227513.1 | - | 1 - 2 | 327 | 67 |
BT004636.1 | - | 1 - 2 | 168 | 67 |