Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PPK1
Entrez Gene ID
844108
Gene Name
putative protein kinase 1
Gene Aliases
AT1G77720, T32E8.5, T32E8_5, putative protein kinase 1
Chromosome Location
Chr.1: 29210496 - 29214135 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_106421.5 NP_565160.3 4 - 5 1837 80
GenBank mRNA AK227513.1 - 4 - 5 1762 80
BT004636.1 - 4 - 5 1603 80

View Full Product Details