Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NIA1
Entrez Gene ID
844112
Gene Name
nitrate reductase 1
Gene Aliases
AT1G77760, GNR1, NITRATE REDUCTASE, NITRATE REDUCTASE 1, NR1, T32E8.9, T32E8_9, nitrate reductase 1
Chromosome Location
Chr.1: 29235721 - 29239484 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_106425.3 NP_177899.1 1 - 2 1167 114
GenBank mRNA AF424624.1 - 1 - 2 1126 114
AY090950.1 - 1 - 2 1128 114
BT000989.1 - 1 - 2 1050 114

View Full Product Details