Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TPPB
Entrez Gene ID
844144
Gene Name
trehalose-6-phosphate phosphatase
Gene Aliases
AT1G78090, ATTPPB, Arabidopsis thaliana trehalose-6-phosphate phosphatase B, T11I11.2, T11I11_2, TREHALOSE-6-PHOSPHATE PHOSPHATASE, trehalose-6-phosphate phosphatase, trehalose-6-phosphate phosphatase B
Chromosome Location
Chr.1: 29373803 - 29376639 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001334818.1 NP_001322316.1 8 - 9 968 74
NM_106458.4 NP_177932.1 8 - 9 1054 74
GenBank mRNA BT002801.1 - 8 - 9 963 74
BT004348.1 - 8 - 9 902 74

View Full Product Details