Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TSD2
Entrez Gene ID
844160
Gene Name
S-adenosyl-L-methionine-dependent methyltransferases superfamily protein
Gene Aliases
AT1G78240, F3F9.21, F3F9_21, OSU1, QUA2, QUASIMODO2, TUMOROUS SHOOT DEVELOPMENT 2
Chromosome Location
Chr.1: 29432829 - 29436674 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161003.2 NP_001154475.1 8 - 9 2190 66
NM_106474.5 NP_177948.3 7 - 8 2305 66

View Full Product Details