Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SRF5
Entrez Gene ID
844238
Gene Name
STRUBBELIG-receptor family 5
Gene Aliases
AT1G78980, STRUBBELIG-receptor family 5, YUP8H12R.40, YUP8H12R_40
Chromosome Location
Chr.1: 29707625 - 29711421 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_106547.3 NP_178019.2 8 - 9 792 77
GenBank mRNA AY518290.1 - 8 - 9 792 77
FJ708683.1 - 8 - 9 637 77

View Full Product Details