The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001198506.1 | NP_001185435.1 | 8 - 9 | 1219 | 122 |
NM_001198507.1 | NP_001185436.1 | 8 - 9 | 1219 | 122 | |
NM_106578.3 | NP_178048.2 | 8 - 9 | 1181 | 122 | |
GenBank mRNA | EF426860.1 | - | 8 - 9 | 1012 | 122 |