Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NUA
Entrez Gene ID
844268
Gene Name
nuclear pore anchor
Gene Aliases
AT1G79280, AtTPR, TRANSLOCATED PROMOTER REGION, YUP8H12R.12, YUP8H12R_12, nuclear pore anchor
Chromosome Location
Chr.1: 29818970 - 29833016 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198506.1 NP_001185435.1 9 - 10 1317 105
NM_001198507.1 NP_001185436.1 9 - 10 1317 105
NM_106578.3 NP_178048.2 9 - 10 1279 105
GenBank mRNA EF426860.1 - 9 - 10 1110 105

View Full Product Details