Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GAPCP-1
Entrez Gene ID
844291
Gene Name
glyceraldehyde-3-phosphate dehydrogenase of plastid 1
Gene Aliases
AT1G79530, T8K14.5, T8K14_5, glyceraldehyde-3-phosphate dehydrogenase of plastid 1
Chromosome Location
Chr.1: 29915902 - 29919408 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_106601.4 NP_178071.1 2 - 3 484 83
GenBank mRNA AF348583.1 - 2 - 3 164 83
AK117920.1 - 2 - 3 310 83

View Full Product Details