Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NUDT3
Entrez Gene ID
844308
Gene Name
nudix hydrolase homolog 3
Gene Aliases
AT1G79690, F20B17.11, F20B17_11, atnudt3, nudix hydrolase homolog 3
Chromosome Location
Chr.1: 29985046 - 29990388 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_106618.5 NP_565218.1 20 - 21 2579 85
GenBank mRNA AY120774.1 - 20 - 21 2295 85
BT000142.1 - 20 - 21 2265 85

View Full Product Details