Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PSF1
Entrez Gene ID
844359
Gene Name
partner of SLD five 1
Gene Aliases
AT1G80190, F18B13.27, F18B13_27, partner of SLD five 1
Chromosome Location
Chr.1: 30158923 - 30161829 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001334963.1 NP_001323117.1 2 - 3 432 71
NM_001334964.1 NP_001323120.1 1 - 2 522 71
NM_001334965.1 NP_001323116.1 1 - 2 508 71
NM_001334966.1 NP_001323119.1 2 - 3 400 71
NM_106668.6 NP_178136.1 2 - 3 415 71
GenBank mRNA AK175552.1 - 2 - 3 365 71
BT010484.1 - 1 - 2 73 71

View Full Product Details