Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- GA3OX2
- Entrez Gene ID
-
844374
- Gene Name
- gibberellin 3-oxidase 2
- Gene Aliases
-
ARABIDOPSIS THALIANA GIBBERELLIN-3-OXIDASE 2, AT1G80340, ATGA3OX2, F5I6.9, F5I6_9, GA4H, GIBBERELLIN 3 BETA-HYDROXYLASE, gibberellin 3-oxidase 2
- Chromosome Location
-
Chr.1: 30200466 - 30202349 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_106683.2 |
NP_178150.1 |
1 - 2 |
656 |
93 |
View Full Product Details