Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ERH3
Entrez Gene ID
844375
Gene Name
P-loop containing nucleoside triphosphate hydrolases superfamily protein
Gene Aliases
AAA1, ARABIDOPSIS THALIANA KATANIN SMALL SUBUNIT, AT1G80350, ATKSS, ATKTN1, BOT1, BOTERO 1, CAD ATPASE, ECTOPIC ROOT HAIR 3, F5I6.10, F5I6_10, FAT ROOT, FRA2, FRAGILE FIBER 2, FRC2, FTR, FURCA2, KATANIN 1, KTN1, LUE1
Chromosome Location
Chr.1: 30205314 - 30208268 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_106684.5 NP_178151.1 2 - 3 819 86
GenBank mRNA AF048706.1 - 2 - 3 610 86
AF358779.1 - 2 - 3 623 86
AK228515.1 - 2 - 3 756 86
AY059919.1 - 2 - 3 756 86
BT000149.1 - 2 - 3 601 86

View Full Product Details