Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IAA15
Entrez Gene ID
844379
Gene Name
indole-3-acetic acid inducible 15
Gene Aliases
AT1G80390, F5I6.14, F5I6_14, indole-3-acetic acid inducible 15
Chromosome Location
Chr.1: 30221629 - 30222769 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001334986.1 NP_001319427.1 2 - 3 659 103

View Full Product Details