Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NHO1
Entrez Gene ID
844385
Gene Name
Actin-like ATPase superfamily protein
Gene Aliases
AT1G80460, GLI1, T21F11.21, T21F11_21, nonhost resistance to P. s. phaseolicola 1
Chromosome Location
Chr.1: 30246683 - 30249337 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_106694.4 NP_178161.1 3 - 4 1601 58
GenBank mRNA AK316784.1 - 3 - 4 1363 58
AY234854.1 - 3 - 4 1363 58

View Full Product Details