Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WRKY66
Entrez Gene ID
844398
Gene Name
WRKY DNA-binding protein 66
Gene Aliases
AT1G80590, ATWRKY66, T21F11.8, T21F11_8, WRKY DNA-binding protein 66
Chromosome Location
Chr.1: 30296166 - 30297224 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_106707.2 NP_178174.1 1 - 2 272 71
GenBank mRNA AB493541.1 - 1 - 2 204 71

View Full Product Details