Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NRAMP1
Entrez Gene ID
844422
Gene Name
natural resistance-associated macrophage protein 1
Gene Aliases
AT1G80830, ATNRAMP1, F23A5.18, F23A5_18, PMIT1, natural resistance-associated macrophage protein 1
Chromosome Location
Chr.1: 30372668 - 30376072 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001335016.1 NP_001321606.1 7 - 8 755 61
NM_106731.3 NP_178198.1 6 - 7 952 61
GenBank mRNA AF165125.1 - 6 - 7 598 61
BT029300.1 - 6 - 7 531 61

View Full Product Details