Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
J8
Entrez Gene ID
844432
Gene Name
Chaperone DnaJ-domain superfamily protein
Gene Aliases
AT1G80920, AtJ8, AtToc12, DJC22, DNA J protein C22, F23A5.28, F23A5_28, Toc12, translocon at the outer envelope membrane of chloroplasts 12
Chromosome Location
Chr.1: 30403503 - 30404713 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_106740.5 NP_178207.1 2 - 3 471 145
GenBank mRNA AF332461.1 - 2 - 3 307 145
AY040017.1 - 2 - 3 443 145
AY079369.1 - 2 - 3 307 145
AY088122.1 - 2 - 3 444 145

View Full Product Details