Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RAPTOR1
Entrez Gene ID
820032
Gene Name
Regulatory-associated protein of TOR 1
Gene Aliases
AT3G08850, ATRAPTOR1B, RAPTOR1B
Chromosome Location
Chr.3: 2686457 - 2695400 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111719.3 NP_566335.1 16 - 17 3517 82
GenBank mRNA AY059838.1 - 16 - 17 3417 82
AY769948.1 - 16 - 17 3417 82
BT008813.1 - 16 - 17 3028 82

View Full Product Details