Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DEX1
Entrez Gene ID
820063
Gene Name
defective in exine formation protein (DEX1)
Gene Aliases
AT3G09090, DEFECTIVE IN EXINE FORMATION 1
Chromosome Location
Chr.3: 2782698 - 2787930 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001125130.1 NP_001118602.1 6 - 7 1529 105
NM_001202915.1 NP_001189844.1 6 - 7 1410 105
NM_111744.5 NP_566343.1 6 - 7 1655 105

View Full Product Details