Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PTAC13
Entrez Gene ID
820077
Gene Name
plastid transcriptionally active 13
Gene Aliases
AT3G09210, plastid transcriptionally active 13
Chromosome Location
Chr.3: 2825142 - 2826922 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111755.3 NP_566346.1 2 - 3 636 75
GenBank mRNA AY049246.1 - 2 - 3 594 75
AY129491.1 - 2 - 3 564 75

View Full Product Details