Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ADK1
Entrez Gene ID
820140
Gene Name
adenosine kinase 1
Gene Aliases
ADENOSINE KINASE 1, AT3G09820, ATADK1, adenosine kinase 1
Chromosome Location
Chr.3: 3011780 - 3014944 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111817.4 NP_187593.1 7 - 8 811 78
NM_202540.1 NP_974269.1 6 - 7 788 78
GenBank mRNA AF180894.1 - 7 - 8 518 78
AF375451.1 - 7 - 8 538 78
BT033101.1 - 5 - 6 343 78

View Full Product Details