Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SCPL49
Entrez Gene ID
820205
Gene Name
SERINE CARBOXYPEPTIDASE-LIKE 49
Gene Aliases
AT3G10410, CARBOXYPEPTIDASE Y, CPY, SERINE CARBOXYPEPTIDASE-LIKE 49
Chromosome Location
Chr.3: 3235290 - 3238267 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111876.5 NP_187652.1 3 - 4 836 138
GenBank mRNA AY091767.1 - 3 - 4 653 138
AY149954.1 - 3 - 4 632 138

View Full Product Details