Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BSMT1
Entrez Gene ID
820321
Gene Name
S-adenosyl-L-methionine-dependent methyltransferases superfamily protein
Gene Aliases
AT3G11480, ATBSMT1
Chromosome Location
Chr.3: 3614421 - 3617342 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111981.5 NP_187755.2 1 - 2 163 99
GenBank mRNA AK221911.1 - 1 - 2 109 99
AY224595.1 - 1 - 2 40 99
BT022049.1 - 1 - 2 104 99

View Full Product Details