Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPY
Entrez Gene ID
820327
Gene Name
Tetratricopeptide repeat (TPR)-like superfamily protein
Gene Aliases
AT3G11540, SPINDLY
Chromosome Location
Chr.3: 3631887 - 3638100 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111987.4 NP_187761.1 5 - 6 1446 87
GenBank mRNA AK221314.1 - 5 - 6 1172 87
KJ138828.1 - 4 - 5 811 87
U62135.1 - 5 - 6 1161 87

View Full Product Details