Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SDH2-1
Entrez Gene ID
822359
Gene Name
succinate dehydrogenase 2-1
Gene Aliases
AT3G27380, succinate dehydrogenase 2-1
Chromosome Location
Chr.3: 10129610 - 10132882 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_113653.4 NP_189374.1 2 - 3 1640 66

View Full Product Details