Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MDAR4
Entrez Gene ID
822402
Gene Name
monodehydroascorbate reductase 4
Gene Aliases
AT3G27820, ATMDAR4, MONODEHYDROASCORBATE REDUCTASE 4, monodehydroascorbate reductase 4
Chromosome Location
Chr.3: 10315010 - 10318154 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_113698.4 NP_189420.1 4 - 5 614 114
GenBank mRNA AY039980.1 - 4 - 5 472 114
AY133800.1 - 4 - 5 375 114

View Full Product Details