Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABCB16
Entrez Gene ID
822465
Gene Name
P-glycoprotein 16
Gene Aliases
AT3G28360, ATP-binding cassette B16, P-glycoprotein 16, PGP16
Chromosome Location
Chr.3: 10611024 - 10616361 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_113756.5 NP_189477.4 6 - 7 2368 83

View Full Product Details