Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ABCB18
Entrez Gene ID
822468
Gene Name
P-glycoprotein 18
Gene Aliases
AT3G28390, ATP-binding cassette B18, P-glycoprotein 18, PGP18
Chromosome Location
Chr.3: 10629425 - 10633967 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_113759.1 NP_189480.1 1 - 2 202 77

View Full Product Details