Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RLP43
Entrez Gene ID
822523
Gene Name
receptor like protein 43
Gene Aliases
AT3G28890, AtRLP43, receptor like protein 43
Chromosome Location
Chr.3: 10895977 - 10898900 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_113810.2 NP_189531.1 1 - 2 2135 87

View Full Product Details