Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EXPA5
Entrez Gene ID
822543
Gene Name
expansin A5
Gene Aliases
ARABIDOPSIS THALIANA EXPANSIN 5, ARABIDOPSIS THALIANA EXPANSIN A5, AT3G29030, ATEXP5, ATEXPA5, ATHEXP ALPHA 1.4, EXP5, EXPANSIN 5, expansin A5
Chromosome Location
Chr.3: 11011262 - 11013451 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_113824.3 NP_189545.1 1 - 2 552 77
GenBank mRNA BT025711.1 - 1 - 2 169 77
U30478.1 - 1 - 2 269 77

View Full Product Details