Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MPK3
Entrez Gene ID
823706
Gene Name
mitogen-activated protein kinase 3
Gene Aliases
AT3G45640, ATMAPK3, ATMPK3, T6D9.4, mitogen-activated protein kinase 3
Chromosome Location
Chr.3: 16756571 - 16758874 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_114433.3 NP_190150.1 2 - 3 624 80
GenBank mRNA AF386961.1 - 2 - 3 425 80
AK226663.1 - - 428 80
BT000007.1 - 2 - 3 277 80

View Full Product Details