Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PAP19
- Entrez Gene ID
-
823755
- Gene Name
- purple acid phosphatase 19
- Gene Aliases
-
AT3G46120, ATPAP19, PURPLE ACID PHOSPHATASE 19, purple acid phosphatase 19
- Chromosome Location
-
Chr.3: 16935921 - 16939766 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_114481.1 |
NP_190198.1 |
4 - 5 |
655 |
79 |
View Full Product Details