Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MEE39
Entrez Gene ID
823778
Gene Name
Leucine-rich repeat protein kinase family protein
Gene Aliases
AT3G46330, maternal effect embryo arrest 39
Chromosome Location
Chr.3: 17020718 - 17025069 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339245.1 NP_001326106.1 7 - 8 1738 83
NM_114500.5 NP_190217.2 8 - 9 1810 83
GenBank mRNA FJ708732.1 - 8 - 9 1625 83

View Full Product Details