Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PHT4;3
Entrez Gene ID
823851
Gene Name
phosphate transporter 4;3
Gene Aliases
AT3G46980, phosphate transporter 4;3
Chromosome Location
Chr.3: 17306599 - 17310068 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035747.2 NP_001030824.1 2 - 3 751 69
NM_001339291.1 NP_001319701.1 2 - 3 714 69
NM_001339292.1 NP_001326351.1 2 - 3 855 69
NM_001339293.1 NP_001326353.1 1 - 2 863 69
NM_001339295.1 NP_001326352.1 2 - 3 740 69
NM_001339296.1 NP_001326350.1 1 - 2 837 69
NM_114565.3 NP_190282.2 2 - 3 759 69
GenBank mRNA BT015413.1 - 2 - 3 686 69

View Full Product Details