Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PEX11B
Entrez Gene ID
823897
Gene Name
peroxin 11B
Gene Aliases
AT3G47430, peroxin 11B
Chromosome Location
Chr.3: 17480521 - 17481800 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_114611.4 NP_190327.1 1 - 2 464 87
GenBank mRNA AJ520107.1 - 1 - 2 187 87
BT002033.1 - 1 - 2 287 87
BT006271.1 - 1 - 2 187 87

View Full Product Details