Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HA4
Entrez Gene ID
823950
Gene Name
H[+]-ATPase 4
Gene Aliases
AHA4, AT3G47950, H(+)-ATPase 4
Chromosome Location
Chr.3: 17692666 - 17697999 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339341.1 NP_001325714.1 15 - 16 2119 135
NM_114664.4 NP_190378.2 15 - 16 2247 135
GenBank mRNA AK118088.1 - 15 - 16 2211 135

View Full Product Details