Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ALDH10A9
Entrez Gene ID
823972
Gene Name
aldehyde dehydrogenase 10A9
Gene Aliases
AT3G48170, BADH, BETAINE ALDEHYDE DEHYDROGENASE, aldehyde dehydrogenase 10A9
Chromosome Location
Chr.3: 17785854 - 17790026 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_114686.4 NP_190400.1 5 - 6 683 75
GenBank mRNA AF370333.1 - 5 - 6 631 75
AY062987.1 - 5 - 6 575 75

View Full Product Details