The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001339379.1 | NP_001325849.1 | 3 - 4 | 607 | 85 |
NM_114737.5 | NP_190447.1 | 3 - 4 | 607 | 85 | |
GenBank mRNA | AB074928.1 | - | 3 - 4 | 312 | 85 |
AY054489.1 | - | 3 - 4 | 518 | 85 | |
AY059882.1 | - | 3 - 4 | 419 | 85 | |
AY114662.1 | - | 3 - 4 | 281 | 85 | |
BT006615.1 | - | 3 - 4 | 281 | 85 |